Learn More
Medchemexpress LLC Chitosan (≥90% deacetylated, Low viscosity,<200mPa.s) | 9012-76-4 | ≥90% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB2144E
Encompass_Preferred
Chitosan (Deacetylated chitin) is a polysaccharide obtained by deacetylating chitin. It exhibits antimicrobial activity against various bacteria and fungi, making it suitable for a variety of applications.
  • Penetrates bacterial cell walls and inhibits DNA transcription and mRNA synthesis.
  • Forms an impermeable layer around cells, changing permeability and blocking transport.
  • Used in water treatment.
  • Suitable for wound-healing materials.
  • Functions as a pharmaceutical excipient or drug carrier.
  • Researched for obesity.
  • Acts as a scaffold for tissue engineering.
Catalog No. 50-004-14403