Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | ≥80% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB2144G10G
Encompass_Preferred
Chitosan (Deacetylated chitin; Poly(D-glucosamine)) is a polysaccharide obtained by deacetylating chitin. It exhibits antimicrobial activity against various bacteria and fungi. Chitosan is recognized as a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. It also demonstrates antitumor, antibacterial, antifungal, and antioxidant activities.
  • Obtained by deacetylating chitin
  • Exhibits antimicrobial activity against various bacteria and fungi
  • Polycationic linear polysaccharide
  • Versatile biomaterial due to non-toxicity, low allergenicity, biocompatibility, and biodegradability
  • Demonstrates antitumor, antibacterial, antifungal, and antioxidant activities
Catalog No. 50-004-11314