Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | ≥85.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB2144K1G
Encompass_Preferred
Chitosan (Deacetylated chitin) (≥85% deacetylated, viscosity>90 mPa.s, MW 15000) is a polysaccharide obtained by deacetylating chitin, and exhibits antimicrobial activity against various bacteria and fungi. It is a polycationic linear polysaccharide with non-toxicity, low allergenicity, biocompatibility, biodegradability, and has antitumor, antibacterial, antifungal, and antioxidant activities.
  • Polycationic linear polysaccharide
  • Non-toxicity
  • Low allergenicity
  • Biocompatibility
  • Biodegradability
  • Antitumor activity
  • Antibacterial activity
  • Antifungal activity
  • Antioxidant activity
Catalog No. 50-004-10447