Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | ≥85% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB2144K
Encompass_Preferred
Chitosan is a polysaccharide obtained by deacetylating chitin, and exhibits antimicrobial activity against various bacteria and fungi. It is a polycationic linear polysaccharide, and a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. Chitosan also possesses antitumor, antibacterial, antifungal, and antioxidant activities.
  • Antimicrobial activity against various bacteria and fungi.
  • Non-toxicity.
  • Low allergenicity.
  • Biocompatibility.
  • Biodegradability.
  • Antitumor activity.
  • Antibacterial activity.
  • Antifungal activity.
  • Antioxidant activity.
Catalog No. 50-004-14471