Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | ≥85% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB2144B5G
Encompass_Preferred
Chitosan (MW 30000), also known as deacetylated chitin, is a polycationic linear polysaccharide with an average molecular weight of 30000. It is recognized as a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. This compound also demonstrates antitumor, antibacterial, antifungal, and antioxidant activities.
  • Exhibits anticancer properties by reducing growth potential and inducing pro-apoptotic effects in certain cell lines.
  • Functions as an antibacterial agent by penetrating bacterial cell walls, inhibiting DNA transcription, and altering cell permeability.
  • Can be utilized in water treatment and wound-healing materials.
  • Serves as a pharmaceutical excipient or drug carrier.
  • Applicable in obesity research and as a scaffold for tissue engineering.
Catalog No. 50-004-11438