Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | ≥90% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB2144J1G
Encompass_Preferred
Chitosan is a polysaccharide obtained by deacetylating chitin, exhibiting antimicrobial activity against various bacteria and fungi. It serves as a versatile biomaterial in drug delivery, tissue engineering, and wound healing. It can also be used in water treatment, as a pharmaceutical excipient or drug carrier, in obesity research, and as a scaffold for tissue engineering.
  • Antimicrobial activity against various bacteria and fungi
  • Versatile biomaterial for drug delivery, tissue engineering, and wound healing
  • Used in water treatment
  • Pharmaceutical excipient or drug carrier
  • Research in obesity
  • Scaffold for tissue engineering
  • Appearance: Solid
  • Color: Off-white to light yellow
Catalog No. 50-004-10918