Learn More
Medchemexpress LLC Chitosan | 9012-76-4 | MFCD00161512 | >=80% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB2144F100G
Encompass_Preferred
Chitosan is a polysaccharide obtained by deacetylating chitin, exhibiting antimicrobial activity against various bacteria and fungi. It is a polycationic linear polysaccharide derived from chitin, known for its versatility as a biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. Chitosan also possesses antitumor, antibacterial, antifungal, and antioxidant activities.
  • Polysaccharide obtained by deacetylating chitin.
  • Exhibits antimicrobial activity against various bacteria and fungi.
  • Polycationic linear polysaccharide.
  • Versatile biomaterial.
  • Non-toxicity, low allergenicity, biocompatibility, and biodegradability.
  • Antitumor, antibacterial, antifungal, and antioxidant activities.
Catalog No. 50-004-10921