Learn More
Medchemexpress LLC Chitosan hydrochloride (80%-90% deacetylated) | 70694-72-3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB2144H
Encompass_Preferred
Chitosan hydrochloride 80%-90% deacetylated is the deacetylated, hydrochloride form of chitosan. It is a polysaccharide obtained by deacetylating chitin and exhibits antimicrobial activity against various bacteria and fungi. This product is utilized in agriculture, water treatment, and pharmaceutical aspects.
  • Deacetylated, hydrochloride form of chitosan
  • Polysaccharide obtained by deacetylating chitin
  • Exhibits antimicrobial activity against various bacteria and fungi
  • Utilized in agriculture, water treatment, and pharmaceutical aspects
  • Inhibits the growth of bacteria and fungi by binding to negatively charged bacterial components and disrupting cell membrane permeability
Catalog No. 50-004-13940