Learn More
Santa Cruz Biotechnology CHK1 (G-4) MONOCLONAL ANTIBODY
Supplier:  Santa Cruz Biotechnology SC8408
Encompass
Chk1 (G-4),mouse monoclonal IgG2a; 200 ug/ml,raised against amino acids 1-476 representing full length Chk1 of human origin,recommended for detection of Chk1 of m,r,h origin by WB, IP, IHC, FCM, agarose conjugate for IP studies, sc-8408 AC
Catalog No. NC9914467