Learn More
Sino Biological Peptide Substrates: Chktide
Supplier:  Sino Biological C1058
Encompass_Preferred
The Chktide peptide sequence (KKKVSRSGLYRSPSMPENLNRPR) is based on the human CDC25C protein isoform A (amino acid 205-225).
Catalog No. 50-256-9622