Learn More
ATCC CHLA-01R-MED; Medulloblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL3034
Encompass
CHLA-01R-MED is a cell line exhibiting rounded morphology that was isolated from the brain of an 8-year-old male with recurrent metastatic medulloblastoma. It was deposited by the Children's Hospital of Los Angeles and can be used for neuroscience research.
Catalog No. 50-238-2158