Learn More
ATCC Chlamydia suis Everett et al., Strain S45 (ATCC VR-1474) - Frozen reference culture
Supplier:  ATCC VR1474
Encompass
Chlamydia suis; Strain S45 (ATCC VR-1474) is the type strain isolated from swine and supplied as a frozen reference culture for laboratory research. It is intended for use under appropriate biosafety conditions for diagnostics, comparative studies, and method development.
  • Type strain S45.
  • Porcine intestinal or fecal isolate.
  • Supplied as a frozen culture for research use.
  • Stored in the vapor phase of liquid nitrogen.
  • Biosafety level 2 organism.
  • Optimal growth at 37°C in a humidified 5% CO2 atmosphere; incubation approximately 72 hours.
  • Used as a reference strain for diagnostics and comparative genomics.
Catalog No. NC3847969