Learn More
ATCC Chlamydia trachomatis H; Strain: UW-43/Cx; DNA
Supplier:  ATCC VR879D
Encompass
Chlamydia DNA, in total DNA, was isolated from McCoy cells (ATCC CRL-1696) infected with Chlamydia trachomatis serovar H strain UW-43/Cx (ATCC VR-879). This product was prepared using methods known to inactivate agents. It is suitable for use in PCR and other molecular biology applications. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4356
 Restricted Item