Learn More
ATCC Chlamydophila pneumoniae; Strain: CM-1; DNA
Supplier:  ATCC VR1360D
Encompass
Chlamydophila DNA, in total DNA, was isolated from Hep-2 cells (ATCC CCL-23) infected with Chlamydophila pneumoniae strain CM-1 (ATCC VR-1360). This product was prepared using methods known to inactivate the infecting agent. It is suitable for use in PCR and other molecular biology applications. The source organism and host cells are also available through the ATCC Catalog.
Catalog No. 50-238-1897