Learn More
PALINTEST USA Kemio Chlorine Dioxide & Chlorite 3 months Refill Kit
Supplier:  PALINTEST USA KEM230CDX
Encompass_Preferred
Kemio™ Chlorine Dioxide & Chlorite Refill Kit includes sensors and reagents for up to 3 months worth of chlorine dioxide and chlorite testing.Kit contents: 300 single-use CDX sensors, 1 x CR-1 reagent, 1 x CR-2 reagent, 1 x glycine reagent, 1 x pack of 10 stirring rods
Catalog No. NC2050107