Learn More
STA PHARMACEUTICAL US LLC Chloracetyl C6 amine amidite | 1 g | CAS 220555-61-3 | InChIKey LHDZYEHTVUMWHK-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10721AK601GM
Chloracetyl C6 amine amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 220555-61-3
- MDL: No data
- InChIKey: LHDZYEHTVUMWHK-UHFFFAOYSA-N
- Molecular Weight: 393.892761998
- Molecular Formula: C17H33ClN3O3P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 6-(2-chloroacetamido)hexyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(C)N(C(C)C)P(OCCCCCCNC(CCl)=O)OCCC#N
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.