Learn More
Medchemexpress LLC Chlorahololide C | 1007859-25-7 | 1 MG

Supplier: Medchemexpress LLC HYN84041MG
Chlorahololide C is a lindenane sesquiterpenoid dimer isolated from *Chloranthus holostegius*. It functions as a potent and selective potassium channel blocker, with an IC50 of 3.6 μM. For research use only. In vitro, Chlorahololide C (0.1-100 μM) inhibits the delayed rectifier K+ current (IK) in rat dissociated hippocampal neurons, with an IC50 of 3.6 μM.
- Molecular weight: 576.63
- Formula: C33H36O9
- Appearance: Solid
- Color: White to off-white
- Purity: 98.0%
- Structure classification: Terpenoids, sesquiterpenes
- Initial source: Plants, Chloranthaceae, *Chloranthus holostegius* (Hand.-Mazz.) Pei et Shan
- Potent and selective potassium channel blocker
- Suitable for research applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.