Learn More
Cayman Chemical ChlorambucIl 250mg
SDP
Supplier:  Cayman Chemical 23744250 MG
Encompass_Preferred
A DNA alkylating agent; cross-links DNA at 1 mg/ml and targets guanine-rich regions; reduces fetal brain size and induces microcephaly when administered to pregnant mice at 8 and 15 mg/kg
Catalog No. 50-299-4979