Learn More
Cayman Chemical ChloramIn-Thydrate 5g
SDP
Supplier:  Cayman Chemical 305335 G
Encompass_Preferred
A common reagent; has been used as a reagent in aminohydroxylation and allylic amination reactions, as a nitrogen source for the aziridination of alkenes and olefins, and in the deprotection of thio groups in sulfur-containing compounds, among others; has been used as a reagent in the synthesis of Factor Xa inhibitors; also an antiseptic agent that is bactericidal against S. epidermidis, S. aureus, E. faecalis, E. coli, P. mirabilis, and E. cloacae at 0.2% w/v
Catalog No. 50-299-1661