Learn More
Selleck Chemical LLC Chloramine-T
SDP
Supplier:  Selleck Chemical LLC S640325MG
Encompass_Preferred
Chloramine-T is a N-chlorinated and N-deprotonated sulfonamide used as a biocide and a mild disinfectant
Catalog No. 50-000-04648