Learn More
Alkali Scientific Chloramphenicol, 25 G
SDP
Supplier:  Alkali Scientific CHL003CP
Encompass_Preferred
Chloramphenicol is a broad-spectrum antibiotic supplied in a 25 gram laboratory format for microbiological and research applications. It inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit, preventing peptide chain formation. This activity provides effective control against both Gram-positive and Gram-negative bacteria. It is commonly used in selective culture media, molecular biology protocols, and antimicrobial testing procedures. The compound is valued for its stability and consistent performance in laboratory environments. It is frequently applied in cloning workflows, plasmid selection, and microbiological research where reliable inhibition of bacterial growth is required.
Catalog No. 50-006-60829