Learn More
Abcam Chloramphenicol, antibiotic agent, 1G
Supplier:  Abcam AB1424111G
Encompass_Preferred
MW 323.13 Da, Purity >99%. Broad spectrum antibiotic agent. Binds selectively to A2451 and A2452 residues in the 23S rRNA of the 50S ribosomal subunit. Prevents peptide bond formation and protein chain elongation inhibiting protein biosynthesis. Shows bacteriostatic effects against a wide variety of Gram-positive and Gram-negative bacteria in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6252