Learn More
Medchemexpress LLC Chloramphenicol-d5 | 202480-68-0 | 99.9% | 1 MG
Supplier: Medchemexpress LLC HYB0239S1MG
Chloramphenicol-d5 is a deuterium-labeled, broad-spectrum antibiotic effective against bacterial infections. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules to serve as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS