Learn More
Gold Biotechnology Inc Chloramphenicol, USP Grade 100 g
SDP
Supplier:  Gold Biotechnology Inc C105100
Encompass_Preferred
Chloramphenicol, also known as chlornitromycin, is a broad-spectrum, bacteriostatic antibiotic derived from Streptomyces venezuelae. It is effective against several gram-positive and gram-negative bacteria and commonly used in researching protein synthesis and to select for chloramphenicol-resistant transformed cells or the bacterial CAT gene.
Catalog No. 50-153-2756