Learn More
Medchemexpress LLC Chlorendic acid | 115-28-6 | 99.8% | 25 G

Supplier: Medchemexpress LLC 25GHY127074
Chlorendic acid (HET acid) is a diacid component utilized in the synthesis of oligoesters. These oligoesters possess potential flame retardant properties, attributed to the degradation by chlorine radicals.
- White to off-white solid
- Molecular weight of 388.84
- Chemical formula C9H4Cl6O4
- Purity of 99.75%
- Soluble in DMSO at concentrations of ≥ 100 mg/mL
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.