Learn More
Medchemexpress LLC Chlorendic acid | 115-28-6 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1270745G
Encompass_Preferred
Chlorendic acid, also known as HET acid, is utilized as a diacid component for the synthesis of oligoesters. It possesses potential flame retardant properties due to degradation by chlorine radicals. This product is intended for research use only.
  • Diacid component for oligoester synthesis
  • Provides potential flame retardant properties
  • Degradation by chlorine radicals
  • For research use only
Catalog No. 50-003-42436