Learn More
Medchemexpress LLC Chlorendic acid (HET acid) | 115-28-6 | 99.8% | 10 G
SDP
Supplier:  Medchemexpress LLC HY12707410G
Encompass_Preferred
Chlorendic acid, also known as HET acid, is utilized as a diacid component for synthesizing oligoesters that exhibit potential flame retardant properties. Its degradation by chlorine radicals contributes to the flame retardancy of HET acid-based oligoesters when combined with aliphatic diols.
  • Used as a diacid component for synthesis
  • Forms oligoesters with potential flame retardant properties
  • Degradation by chlorine radicals contributes to flame retardancy
Catalog No. 50-003-40042