Learn More
Medchemexpress LLC Chlorfenapyr | 122453-73-0 | 99.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB084010MM/1ML
Encompass_Preferred
Chlorfenapyr is a pyrrole insecticide that acts as a pro-insecticide. Its active metabolite, CL 303268, uncouples oxidative phosphorylation in mitochondria, disrupting ATP production. This leads to cell dysfunction and organism death. Chlorfenapyr is utilized for termite control and crop protection against various insect and mite pests.
  • Functions as a pro-insecticide
  • Active metabolite uncouples oxidative phosphorylation in mitochondria
  • Disrupts ATP production
  • Leads to cell dysfunction and organism death
  • Exhibits low mammalian toxicity
  • Used for termite control
  • Used for crop protection against various insect and mite pests
  • For research use only
Catalog No. 50-004-08850