Learn More
Medchemexpress LLC Chlorfenapyr | 122453-73-0 | 99.4% | 407.61 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYB0840
Encompass_Preferred
Chlorfenapyr is a pyrrole insecticide that acts as a pro-insecticide. It is metabolized into an active form, CL 303268, which uncouples oxidative phosphorylation in mitochondria, leading to cell dysfunction and death in target organisms, while exhibiting low mammalian toxicity.
  • Functions as a pro-insecticide
  • Active metabolite uncouples oxidative phosphorylation
  • Disrupts ATP production in target organisms
  • Low mammalian toxicity
  • Effective against mites
  • Used for termite control
  • Protects crops against various insect and mite pests
Catalog No. 50-003-65021