Learn More
Medchemexpress LLC Chlorfenapyr | 122453-73-0 | 99.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB084050MG
Encompass_Preferred
Chlorfenapyr is a pyrrole insecticide that functions as a pro-insecticide. Its active metabolite, CL 303268, is formed when mixed function oxidase oxidizes and removes the N-ethoxymethyl group. This active form uncouples oxidative phosphorylation in mitochondria, disrupting ATP production and leading to cell dysfunction and death of the organism. Chlorfenapyr is utilized for termite control and crop protection against various insect and mite pests. It has low mammalian toxicity.
  • Pyrrole insecticide.
  • Mode of action involves oxidation by mixed function oxidase to form active metabolite CL 303268.
  • CL 303268 uncouples oxidative phosphorylation, leading to disruption of ATP production and energy loss.
  • Used for termite control and crop protection against insect and mite pests.
  • Low mammalian toxicity.
Catalog No. 50-004-08819