Learn More
Medchemexpress LLC Chlorfenapyr | 122453-73-0 | 99.6% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0840500MG
Encompass_Preferred
Chlorfenapyr is a pyrrole insecticide that functions as a pro-insecticide. Its mode of action involves oxidative removal of its N-ethoxymethyl group to form the active metabolite, CL 303268, which uncouples oxidative phosphorylation in mitochondria, leading to cell dysfunction. It is primarily used for termite control and crop protection against various insect and mite pests, and exhibits low mammalian toxicity.
  • Pyrrole insecticide
  • Functions as a pro-insecticide
  • Active metabolite CL 303268
  • Used for termite control
  • Protects crops against insect and mite pests
  • Uncouples oxidative phosphorylation
  • Low mammalian toxicity
  • Off-white to light yellow solid appearance
Catalog No. 50-004-07634