Learn More
Medchemexpress LLC Chlorfenapyr | 122453-73-0 | MFCD01631152 | 99.4% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0840
Encompass_Preferred
Chlorfenapyr is a pyrrole insecticide that acts as a pro-insecticide. Its mode of action involves mixed-function oxidase oxidizing and removing the Nethoxymethyl group to form the active metabolite, CL 303268. This active metabolite uncouples oxidative phosphorylation in the mitochondria, disrupting ATP production and leading to cell dysfunction and organism death.
  • Pyrrole insecticide
  • Used for termite control
  • Used for crop protection against various insect and mite pests
  • Low mammalian toxicity
Catalog No. 50-004-07066