Learn More
Medchemexpress LLC Chlorfenapyr (Standard) | 122453-73-0 | 99.6% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0840
Encompass_Preferred
Chlorfenapyr (Standard) is an analytical standard and pyrrole insecticide. It is intended for research and analytical applications, functioning by oxidation to form the active metabolite, CL 303268. This product is utilized for termite control and crop protection against various insect and mite pests. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Functions as a pyrrole insecticide
  • Oxidizes to form active metabolite, CL 303268
  • Used for termite control and crop protection
  • Utilized in HPLC, GC, and MS experiments
Catalog No. 50-004-50672