Learn More
Medchemexpress LLC Chlorhexidine-d8 dihydrochloride | 2012598-75-1 | 95.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1145S1MG
Encompass_Preferred
Chlorhexidine-d8 dihydrochloride is a deuterium labeled form of Chlorhexidine dihydrochloride, an orally active cationic antimicrobial agent. It targets microbial cell membranes, binding to phospholipids non-specifically to destroy membrane structure and induce leakage of cell contents. It exhibits broad-spectrum bactericidal activity against both Gram-positive and Gram-negative bacteria by interfering with membrane permeability, causing protein precipitation, and disrupting energy metabolism. This leads to rapid inhibition of microbial growth and induction of cell death.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13342