Learn More
Medchemexpress LLC HY-W013699 100mg Medchemexpress, Chlorhexidine (diacetate) CAS:56-95-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW013699100MG
Encompass_Preferred
Medchemexpress, HY-W013699 100mg Chlorhexidine (diacetate) CAS:56-95-1 Chlorhexidine diacetate is a biguanide disinfectant with rapid bactericidal activity against both Gram-positive and Gram-negative organism. The antibacterial effect of chlorhexidine diacetate is related to its action on the bacterial cell membrane and to precipitation of intracellular contents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3351