Learn More
Apexbio Technology LLC Chlorhexidine digluconate 18472-51-0 50mg
SDP
Supplier:  Apexbio Technology LLC B352550
Encompass_Preferred
Chlorhexidine digluconate (18472-51-0) is a small-molecule antimicrobial agent It exerts its biological activity primarily through perturbation of bacterial cell membrane integrity causing disruption of membrane function and subsequent bacterial cell death In typical microbiological assays chlorhexidine digluconate demonstrates inhibitory activity with reported IC50 values generally ranging from 1 to 10 g/mL against several bacterial species Based on these pharmacological properties chlorhexidine digluconate holds research potential in infection control antimicrobial sensitivity assays microbial pathogenesis and cellular membrane permeability studies
Catalog No. 50-000-89176