Learn More
Cayman Chemical ChlorhexIdInhydrochlorIde 10g
SDP
Supplier:  Cayman Chemical 1734310 G
Encompass_Preferred
A bis(biguanide) antimicrobial disinfectant and antiseptic agent; inhibits growth of clinical MRSA isolates (MIC90 = 4 μg/ml); inhibits growth of canine MRSA, MSSA, MRSP, and MSSP isolates (MIC90s = 4, 2, 2, and 1 mg/l, respectively); inhibits growth of E. faecium strains (MICs = 1.2-19.6 μg/ml) and of C. albicans (MIC = 5.15 μg/ml); inhibits MMP-2 and MMP-9 at 0.0001 and 0.002%, respectively
Catalog No. 50-296-6878