Learn More
Hach Company Chloride 2 Indicator Powder Pillows, pk/100
Supplier:  Hach Company 104399
Encompass
For determination of chloride by argentometric titration method with silver nitrate. For test kits. Pack of 100 Powder Pillows.
Catalog No. 50-199-4500