Learn More
Medchemexpress LLC Chlorin e6 (trisodium) | 72984-36-2 | 98.7% | 662.62 g/mol | C34H33N4Na3O6 | 10mg
SDP
Supplier:  Medchemexpress LLC HYW85465910MG
Encompass_Preferred
Chlorin e6 Ce6 (trisodium) is a water-soluble derivative of chlorophyll belonging to the chlorin class of photosensitizers with an absorption wavelength range of 600-670 nm Chlorin e6 trisodium emits characteristic red fluorescence upon light excitation enabling real-time identification of tumor boundaries and progression Chlorin e6 trisodium can be used for the study of photodynamic therapy (PDT) of cancers (bladder cancer) and fluorescence diagnosis of neoplastic lesions[1][2][3]
Catalog No. 50-002-70323