Learn More
Medchemexpress LLC Chlorindanol | 145-94-8 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB099910MM/1ML
Encompass_Preferred
Chlorindanol (7-Chloro-4-indanol) is a topical antiseptic or sanitizer. It is rapidly lethal to vegetative bacteria, Trichophyton sp., C. albicans, E. histolytica cysts and trophozoites, T. vaginalis, and spermatozoa in vitro. Chlorindanol is noted for its low systemic toxicity, good skin/eyes/genital mucosa tolerance, and nonallergenic properties. It is for research use only.
  • Topical antiseptic or sanitizer
  • Rapidly lethal to vegetative bacteria
  • Effective against Trichophyton sp. and C. albicans
  • Effective against E. histolytica cysts and trophozoites
  • Effective against T. vaginalis and spermatozoa in vitro
  • Low systemic toxicity
  • Good skin, eyes, and genital mucosa tolerance
  • Nonallergenic properties
  • For research use only
Catalog No. 50-004-07499