Learn More
Medchemexpress LLC Chlorindanol | 145-94-8 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0999100MG
Encompass_Preferred
Chlorindanol (7-Chloro-4-indanol) is a topical antiseptic or sanitizer. It is rapidly lethal in vitro to vegetative bacteria, Trichophyton sp., C. albicans, E. histolytica cysts and trophozoites, T. vaginalis, and spermatozoa. Chlorindanol has low systemic toxicity, good skin/eyes/genital mucosa tolerance, and is nonallergenic.
  • Topical antiseptic or sanitizer
  • Rapidly lethal to various microorganisms in vitro
  • Low systemic toxicity
  • Good skin, eyes, and genital mucosa tolerance
  • Nonallergenic
Catalog No. 50-004-07535