Learn More
Andwin Scientific Ultra High Range Chlorine Reagents, 300 tests
SDP
Supplier:  Andwin Scientific HI9577103
Encompass
The HI95771-03 are reagents for the colorimetric determination of chlorine in the ultra high range. There are enough reagents for 300 tests to be used with Hanna's HI96771 portable photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.
Catalog No. NC0931721