Learn More
Cayman Chemical ChlormadInon Acetate 1g
SDP
Supplier:  Cayman Chemical 293921 G
Encompass_Preferred
A synthetic progestin; binds to progesterone, androgen, and glucocorticoid receptors in vitro (Kis = 2.5, 3.8, and 16 nM, respectively, for the human receptors); increases the number of endometrial glands and uterine weight in β-estradiol-primed rabbits at 45 UG/kg per day for five days; reduces testosterone-induced increases in the seminal vesicle weight of castrated male rats at 4.6 and 21.5 mg/kg per day for eight days
Catalog No. 50-298-8875