Learn More
Selleck Chemical LLC Chlormadinone acetate S4593-100mg
SDP
Supplier:  Selleck Chemical LLC S4593100MG
Encompass_Preferred
Chlormadinone acetate (Gestafortin) is a steroidal progestin with additional antiandrogen and antigonadotropic effects used often in combinations as an oral contraceptive Its half-life is about 34-38 hours
Catalog No. 50-310-8675