Learn More
Medchemexpress LLC Chlormethiazole hydrochloride | 6001-74-7 | 99.9% | C6H9Cl2NS | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA029610MM/1ML
Encompass_Preferred
Chlormethiazole hydrochloride is a potent and orally active GABAA agonist. It inhibits cytochrome P450 isoforms: CYP2A6 and CYP2E1 in human liver microsomes, making it suitable for various research applications.
  • Potent and orally active GABAA agonist
  • Inhibits cytochrome P450 isoforms: CYP2A6 and CYP2E1
  • Anticonvulsant agent
  • Potential for treating convulsive status epilepticus
Catalog No. 50-004-32271