Learn More
Medchemexpress LLC Chlormethiazole hydrochloride | 6001-74-7 | MFCD00673948 | 99.9% | C6H9Cl2NS | 200 MG
SDP
Supplier:  Medchemexpress LLC HYA0296200MG
Encompass_Preferred
Chlormethiazole hydrochloride is a potent and orally active GABAA agonist, known for its ability to inhibit cytochrome P450 isoforms CYP2A6 and CYP2E1 in human liver microsomes. This compound acts as an anticonvulsant agent, showing potential for the treatment of convulsive status epilepticus. It is provided as a light yellow to yellow solid.
  • Potent and orally active GABAA agonist
  • Inhibits cytochrome P450 isoforms CYP2A6 and CYP2E1
  • Anticonvulsant agent
  • Soluble in DMSO and H2O
Catalog No. 50-004-32911