Learn More
Medchemexpress LLC Chlormethiazole hydrochloride | 6001-74-7 | MFCD00673948 | 99.94% | C6H9Cl2NS | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA029650MG
Encompass_Preferred
Chlormethiazole hydrochloride is a potent and orally active GABAA agonist. It functions by inhibiting cytochrome P450 isoforms CYP2A6 and CYP2E1 in human liver microsomes. This compound acts as an anticonvulsant agent and shows potential for treating convulsive status epilepticus.
  • Potent and orally active GABAA agonist
  • Inhibits cytochrome P450 isoforms (CYP2A6 and CYP2E1)
  • Acts as an anticonvulsant agent
  • Demonstrates potential for treating convulsive status epilepticus
Catalog No. 50-004-32224