Learn More
Apexbio Technology LLC Chlormezanone 80-77-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19095.1
Encompass_Preferred
Chlormezanone (CAS 80-77-3) is a centrally acting small molecule that modulates neurotransmission by potentiating gamma-aminobutyric acid (GABA)-mediated signaling thereby reducing excitability within the central nervous system Its mechanism is thought to parallel that of benzodiazepines primarily facilitating anxiolytic and muscle relaxant effects through enhancement of inhibitory synaptic activity In biomedical research chlormezanone serves as a reference compound for investigating the pharmacology of central nervous system depressants and is frequently used in the identification and development of novel agents targeting anxiety and muscle spasticity
Catalog No. 50-000-92722