Learn More
Strem, An Ascensus Company CAS# 92390-26-6. 250mg. Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), 98%. MFCD07369035
SDP
Supplier:  Strem, An Ascensus Company 440113250MG
Encompass
CAS# 92390-26-6. 250mg. Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), 98%. MFCD07369035. Molecular Weight: 379.93. Molecular Formula: RuCl(C8H12)(C10H15). Color/form: brown microxtls. Strem# 44-0113. http://www.strem.com/catalog/v/44-0113/
Catalog No. 50-901-11793