Learn More
Medchemexpress LLC Chloro-1,5-cyclooctadiene iridium(I) dimer | 12112-67-3 | 671.70 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW008260
Encompass_Preferred
This biochemical reagent is suitable for life science related research. It serves as a precursor to various iridium complexes and is applied in homogeneous catalysis for reactions including carbonylation, hydrosilylation, hydroformylation, asymmetric allylic substitutions, metatheses, and chiral catalysis. Additionally, it plays a role in preparing Crabtree's catalyst for hydrogenation and hydrogen-transfer reactions.
  • Precursor to other iridium complexes
  • Applied in homogeneous catalysis for reactions such as carbonylation, hydrosilylation, hydroformylation, asymmetric allylic substitutions, metatheses, and chiral catalysis
  • Involved in the preparation of Crabtree's catalyst
  • Used for hydrogenation and hydrogen-transfer reactions
Catalog No. 50-004-49899